Product Details

SNP ID
rs184844324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30105229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGTTTTCATCTTTTTTCCTATC[A/G]GCCTGGAGTTGGTCTGGGGAATAAA
Phenotype
MIM: 609316
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM31 PubMed Links

Gene Details

Gene
TRIM31
Gene Name
tripartite motif containing 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007028.4 1007 Silent Mutation GCC,GCT A299A NP_008959.3
XM_011514264.1 1007 Silent Mutation GCC,GCT A298A XP_011512566.1
XM_011514265.1 1007 Silent Mutation GCC,GCT A299A XP_011512567.1
XM_011514266.1 1007 Intron XP_011512568.1
Gene
TRIM31-AS1
Gene Name
TRIM31 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details