Product Details

SNP ID
rs184559948
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:88195646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTTATATATTTTAACCATATTC[A/G]TCTCCATTAAATTTAAAATAACTAT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TMEM161B PubMed Links

Gene Details

Gene
TMEM161B
Gene Name
transmembrane protein 161B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289007.1 1972 Intron NP_001275936.1
NM_001289008.1 1972 UTR 3 NP_001275937.1
NM_153354.4 1972 Intron NP_699185.1
XM_006714553.1 1972 Intron XP_006714616.1
XM_006714554.3 1972 Intron XP_006714617.1
XM_006714555.3 1972 Intron XP_006714618.1
XM_006714556.3 1972 Intron XP_006714619.2
XM_011543201.2 1972 Intron XP_011541503.1
XM_011543202.2 1972 Intron XP_011541504.2
XM_011543203.2 1972 Intron XP_011541505.2
XM_011543204.2 1972 Intron XP_011541506.2
XM_017009093.1 1972 Intron XP_016864582.1
XM_017009094.1 1972 Intron XP_016864583.1
XM_017009095.1 1972 Intron XP_016864584.1
XM_017009096.1 1972 Intron XP_016864585.1
XM_017009097.1 1972 Intron XP_016864586.1
XM_017009098.1 1972 Intron XP_016864587.1
XM_017009099.1 1972 Intron XP_016864588.1
XM_017009100.1 1972 Intron XP_016864589.1
XM_017009101.1 1972 Intron XP_016864590.1
XM_017009102.1 1972 Intron XP_016864591.1
XM_017009103.1 1972 Intron XP_016864592.1
XM_017009104.1 1972 Intron XP_016864593.1
XM_017009105.1 1972 Intron XP_016864594.1
XM_017009106.1 1972 Intron XP_016864595.1
XM_017009107.1 1972 Intron XP_016864596.1
XM_017009108.1 1972 Intron XP_016864597.1
XM_017009109.1 1972 UTR 3 XP_016864598.1
XM_017009110.1 1972 UTR 3 XP_016864599.1

View Full Product Details