Product Details

SNP ID
rs185727260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99875597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGCCATTAGAAATGCTTTTAT[T/A]TATAAAAGAACTACTTAAATATAAA
Phenotype
MIM: 611282
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
DNMBP PubMed Links

Gene Details

Gene
DNMBP
Gene Name
dynamin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318326.1 6280 UTR 3 NP_001305255.1
NM_001318327.1 6280 UTR 3 NP_001305256.1
NM_015221.3 6280 UTR 3 NP_056036.1
XM_006717735.2 6280 UTR 3 XP_006717798.1
XM_011539559.2 6280 UTR 3 XP_011537861.1

View Full Product Details