Product Details

SNP ID
rs185929278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4882128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCTGTTGATCTATGTCTGACC[A/G]TTACGACCCTTCCCACTGTGCTTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51T1 PubMed Links

Gene Details

Gene
OR51T1
Gene Name
olfactory receptor family 51 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004759.1 310 Missense Mutation ATT,GTT I104V NP_001004759.1

View Full Product Details