Product Details

SNP ID
rs186678448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:88195702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTACTGTAATATACAGTAGC[T/A]ATCTCTTCCTTTCTGTTGGATTTTA
Phenotype
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
TMEM161B PubMed Links

Gene Details

Gene
TMEM161B
Gene Name
transmembrane protein 161B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289007.1 1916 Intron NP_001275936.1
NM_001289008.1 1916 UTR 3 NP_001275937.1
NM_153354.4 1916 Intron NP_699185.1
XM_006714553.1 1916 Intron XP_006714616.1
XM_006714554.3 1916 Intron XP_006714617.1
XM_006714555.3 1916 Intron XP_006714618.1
XM_006714556.3 1916 Intron XP_006714619.2
XM_011543201.2 1916 Intron XP_011541503.1
XM_011543202.2 1916 Intron XP_011541504.2
XM_011543203.2 1916 Intron XP_011541505.2
XM_011543204.2 1916 Intron XP_011541506.2
XM_017009093.1 1916 Intron XP_016864582.1
XM_017009094.1 1916 Intron XP_016864583.1
XM_017009095.1 1916 Intron XP_016864584.1
XM_017009096.1 1916 Intron XP_016864585.1
XM_017009097.1 1916 Intron XP_016864586.1
XM_017009098.1 1916 Intron XP_016864587.1
XM_017009099.1 1916 Intron XP_016864588.1
XM_017009100.1 1916 Intron XP_016864589.1
XM_017009101.1 1916 Intron XP_016864590.1
XM_017009102.1 1916 Intron XP_016864591.1
XM_017009103.1 1916 Intron XP_016864592.1
XM_017009104.1 1916 Intron XP_016864593.1
XM_017009105.1 1916 Intron XP_016864594.1
XM_017009106.1 1916 Intron XP_016864595.1
XM_017009107.1 1916 Intron XP_016864596.1
XM_017009108.1 1916 Intron XP_016864597.1
XM_017009109.1 1916 UTR 3 XP_016864598.1
XM_017009110.1 1916 UTR 3 XP_016864599.1

View Full Product Details