Product Details

SNP ID
rs186472801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55665281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCATGTATGTAGCCACAGTGCTG[G/T]AAAATCTACTTATTGTGGTAACTAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4C6 PubMed Links

Gene Details

Gene
OR4C6
Gene Name
olfactory receptor family 4 subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004704.1 115 Nonsense Mutation GAA,TAA E39* NP_001004704.1

View Full Product Details