Product Details

SNP ID
rs186819833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65003000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAGTCCAGATAATTCCT[T/C]GTAAATAAGGTTTTTAATAAATTAC
Phenotype
MIM: 611766
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MTFMT PubMed Links

Gene Details

Gene
MTFMT
Gene Name
mitochondrial methionyl-tRNA formyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139242.3 1400 UTR 3 NP_640335.2
XM_005254158.4 1400 UTR 3 XP_005254215.2

View Full Product Details