Product Details

SNP ID
rs186836076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35118455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTCTGTTCTTTGGTCTCCCCA[T/C]CTGTAAAATATGAGCATTATGTATA
Phenotype
MIM: 610302
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
EDEM2 PubMed Links

Gene Details

Gene
EDEM2
Gene Name
ER degradation enhancing alpha-mannosidase like protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145025.1 Intron NP_001138497.1
NM_018217.2 Intron NP_060687.2

View Full Product Details