Product Details

SNP ID
rs187237351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46500690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGTTTCTCTCCTGTATGAGTT[C/T]GTTGATGTTTAATCAGAGTTGACTT
Phenotype
MIM: 300573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF674 PubMed Links

Gene Details

Gene
ZNF674
Gene Name
zinc finger protein 674
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039891.2 1297 Missense Mutation CAA,CGA Q300R NP_001034980.1
NM_001146291.1 1297 Missense Mutation CAA,CGA Q294R NP_001139763.1
NM_001190417.1 1297 Missense Mutation CAA,CGA Q295R NP_001177346.1
XM_011543941.2 1297 Intron XP_011542243.1
XM_011543943.2 1297 Intron XP_011542245.1
XM_011543944.2 1297 Intron XP_011542246.1
XM_011543945.2 1297 Intron XP_011542247.1
XM_017029728.1 1297 Intron XP_016885217.1
XM_017029729.1 1297 Intron XP_016885218.1
XM_017029730.1 1297 Missense Mutation CAA,CGA Q233R XP_016885219.1
XM_017029731.1 1297 Intron XP_016885220.1
XM_017029732.1 1297 Intron XP_016885221.1
XM_017029733.1 1297 Intron XP_016885222.1

View Full Product Details