Product Details

SNP ID
rs187010424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98734250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTTTCCTTCCTTCTTTCCTTT[C/T]TCTTCTTTTATCTCCAAGGTTAATC
Phenotype
MIM: 607815 MIM: 602233
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 5367 Intron NP_001190994.1
NM_001204066.1 5367 Intron NP_001190995.1
NM_001204067.1 5367 Intron NP_001190996.1
NM_001204068.1 5367 Intron NP_001190997.1
NM_001204069.1 5367 Intron NP_001190998.1
NM_001204070.1 5367 Intron NP_001190999.1
NM_001204079.1 5367 Intron NP_001191008.1
NM_001204080.1 5367 Intron NP_001191009.1
NM_001204081.1 5367 Intron NP_001191010.1
NM_020140.3 5367 Intron NP_064525.1
NM_152788.4 5367 Intron NP_690001.3
NM_181670.3 5367 Intron NP_858056.2
XM_005269028.4 5367 Intron XP_005269085.1
XM_005269029.4 5367 Intron XP_005269086.1
XM_005269032.3 5367 Intron XP_005269089.1
XM_006719504.3 5367 Intron XP_006719567.1
XM_006719505.3 5367 Intron XP_006719568.1
XM_006719506.3 5367 Intron XP_006719569.1
XM_006719507.3 5367 UTR 3 XP_006719570.1
XM_006719508.3 5367 Intron XP_006719571.1
XM_006719509.3 5367 Intron XP_006719572.1
XM_006719510.3 5367 Intron XP_006719573.1
XM_006719512.3 5367 Intron XP_006719575.1
XM_006719513.3 5367 Intron XP_006719576.1
XM_006719514.3 5367 UTR 3 XP_006719577.1
XM_011538571.2 5367 Intron XP_011536873.1
XM_017019651.1 5367 Intron XP_016875140.1
XM_017019652.1 5367 Intron XP_016875141.1
XM_017019653.1 5367 UTR 3 XP_016875142.1
XM_017019654.1 5367 UTR 3 XP_016875143.1
XM_017019655.1 5367 Intron XP_016875144.1
XM_017019656.1 5367 Intron XP_016875145.1
XM_017019657.1 5367 Intron XP_016875146.1
XM_017019658.1 5367 UTR 3 XP_016875147.1
XM_017019659.1 5367 Intron XP_016875148.1
XM_017019660.1 5367 Intron XP_016875149.1
XM_017019661.1 5367 Intron XP_016875150.1
XM_017019662.1 5367 Intron XP_016875151.1
XM_017019663.1 5367 Intron XP_016875152.1
XM_017019664.1 5367 Intron XP_016875153.1
XM_017019665.1 5367 Intron XP_016875154.1
Gene
APAF1
Gene Name
apoptotic peptidase activating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160.2 5367 UTR 3 NP_001151.1
NM_013229.2 5367 UTR 3 NP_037361.1
NM_181861.1 5367 UTR 3 NP_863651.1
NM_181868.1 5367 UTR 3 NP_863658.1
NM_181869.1 5367 UTR 3 NP_863659.1
XM_017019250.1 5367 Intron XP_016874739.1

View Full Product Details