Product Details

SNP ID
rs186729510
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101553147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTCTGGGGCCAAACTTCAGGGT[G/T]ATTCAGAGGTCAAGCCTGAGGTGAG
Phenotype
MIM: 300362
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ARMCX1 PubMed Links

Gene Details

Gene
ARMCX1
Gene Name
armadillo repeat containing, X-linked 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016608.1 588 Missense Mutation GAT,TAT D73Y NP_057692.1

View Full Product Details