Product Details

SNP ID
rs187722336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187670212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAACTGACGGAGTCTGGGGTCC[A/G]AGGGAGCGCCGGTGACACAGCCCAG
Phenotype
MIM: 182450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SST PubMed Links

Gene Details

Gene
SST
Gene Name
somatostatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048.3 202 Missense Mutation NP_001039.1

View Full Product Details