Product Details

SNP ID
rs187399098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57409261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACCACAGCCGACAAGGCCATCA[C/G]CAGCCCAAAGAGCTTGCCAAAGTGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC43A3 PubMed Links

Gene Details

Gene
SLC43A3
Gene Name
solute carrier family 43 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278201.1 1617 Missense Mutation CTG,GTG L429V NP_001265130.1
NM_001278206.1 1617 Missense Mutation CTG,GTG L442V NP_001265135.1
NM_014096.3 1617 Missense Mutation CTG,GTG L429V NP_054815.2
NM_017611.2 1617 Missense Mutation CTG,GTG L429V NP_060081.1
NM_199329.2 1617 Missense Mutation CTG,GTG L429V NP_955361.1

View Full Product Details