Product Details

SNP ID
rs188188728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:616632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCATTAGGGAAAGTGCTGACAAG[C/T]CGCAAGGGATCCCTTGATGGTTCTG
Phenotype
MIM: 606839 MIM: 605047 MIM: 611780 MIM: 182099
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDHR5 PubMed Links

Gene Details

Gene
CDHR5
Gene Name
cadherin related family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171968.1 2989 UTR 3 NP_001165439.1
NM_021924.4 2989 UTR 3 NP_068743.2
NM_031264.3 2989 UTR 3 NP_112554.2
XM_006718253.3 2989 UTR 3 XP_006718316.1
XM_011520188.2 2989 UTR 3 XP_011518490.1
XM_011520189.2 2989 UTR 3 XP_011518491.1
XM_011520190.2 2989 UTR 3 XP_011518492.1
XM_011520191.2 2989 Intron XP_011518493.1
Gene
IRF7
Gene Name
interferon regulatory factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001572.3 2989 Intron NP_001563.2
NM_004029.2 2989 Intron NP_004020.1
NM_004031.2 2989 Intron NP_004022.2
XM_005252906.3 2989 Intron XP_005252963.1
XM_005252907.3 2989 Intron XP_005252964.1
XM_005252909.3 2989 Intron XP_005252966.1
XM_011520066.2 2989 Intron XP_011518368.1
XM_017017674.1 2989 Intron XP_016873163.1
Gene
PHRF1
Gene Name
PHD and ring finger domains 1
There are no transcripts associated with this gene.

Gene
SCT
Gene Name
secretin
There are no transcripts associated with this gene.

View Full Product Details