Product Details

SNP ID
rs188122754
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:37537022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGCTCCATGGAGAAATCAGGAA[T/C]TCTTTCACAAAGAACAGATCCACAG
Phenotype
MIM: 608241
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SNIP1 PubMed Links

Gene Details

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024700.3 2032 UTR 3 NP_078976.2

View Full Product Details