Product Details

SNP ID
rs188707778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50268975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCTGTCCTCCCAACAATTTTC[T/C]GAATTCTATATATTCAGCATTAAAT
Phenotype
MIM: 614759 MIM: 156535
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
There are no transcripts associated with this gene.

Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204136.1 2852 UTR 3 NP_001191065.1
NM_001204137.1 2852 Intron NP_001191066.1
NM_001204138.1 2852 Intron NP_001191067.1
NM_001204139.1 2852 Intron NP_001191068.1
NM_001204140.1 2852 Intron NP_001191069.1
NM_001204141.1 2852 Intron NP_001191070.1
NM_001204142.1 2852 UTR 3 NP_001191071.1
NM_001204143.1 2852 UTR 3 NP_001191072.1
NM_001204151.2 2852 UTR 3 NP_001191080.1
NM_001323942.1 2852 UTR 3 NP_001310871.1
NM_001323947.1 2852 UTR 3 NP_001310876.1
NM_001323949.1 2852 UTR 3 NP_001310878.1
NM_001323950.1 2852 UTR 3 NP_001310879.1
NM_001323951.1 2852 Intron NP_001310880.1
NM_001323952.1 2852 UTR 3 NP_001310881.1
NM_001323953.1 2852 Intron NP_001310882.1
NM_001323954.1 2852 UTR 3 NP_001310883.1
NM_002384.2 2852 UTR 3 NP_002375.1
NM_015844.2 2852 UTR 3 NP_056669.2
NM_015845.3 2852 UTR 3 NP_056670.2
NM_015846.3 2852 UTR 3 NP_056671.2
NM_015847.3 2852 UTR 3 NP_056723.2
XM_005258271.2 2852 UTR 3 XP_005258328.1
XM_006722456.2 2852 UTR 3 XP_006722519.1
XM_011525991.1 2852 UTR 3 XP_011524293.1
XM_011525993.2 2852 UTR 3 XP_011524295.1
XM_011525994.2 2852 UTR 3 XP_011524296.1
XM_011525998.2 2852 Intron XP_011524300.1
XM_011525999.1 2852 UTR 3 XP_011524301.1
XM_011526001.1 2852 UTR 3 XP_011524303.1
XM_011526002.1 2852 UTR 3 XP_011524304.1
XM_011526003.2 2852 UTR 3 XP_011524305.1
XM_011526006.1 2852 UTR 3 XP_011524308.1
XM_011526007.1 2852 UTR 3 XP_011524309.1
XM_017025751.1 2852 UTR 3 XP_016881240.1
XM_017025752.1 2852 UTR 3 XP_016881241.1
XM_017025753.1 2852 UTR 3 XP_016881242.1
XM_017025754.1 2852 Intron XP_016881243.1
XM_017025755.1 2852 Intron XP_016881244.1
XM_017025756.1 2852 UTR 3 XP_016881245.1
XM_017025757.1 2852 UTR 3 XP_016881246.1
XM_017025758.1 2852 UTR 3 XP_016881247.1
XM_017025759.1 2852 UTR 3 XP_016881248.1
XM_017025760.1 2852 UTR 3 XP_016881249.1
XM_017025761.1 2852 Intron XP_016881250.1
XM_017025762.1 2852 UTR 3 XP_016881251.1
XM_017025763.1 2852 UTR 3 XP_016881252.1
XM_017025764.1 2852 UTR 3 XP_016881253.1
XM_017025765.1 2852 UTR 3 XP_016881254.1
XM_017025766.1 2852 UTR 3 XP_016881255.1
XM_017025767.1 2852 UTR 3 XP_016881256.1
XM_017025768.1 2852 Intron XP_016881257.1
XM_017025769.1 2852 UTR 3 XP_016881258.1
XM_017025770.1 2852 UTR 3 XP_016881259.1
XM_017025771.1 2852 UTR 3 XP_016881260.1
XM_017025772.1 2852 Intron XP_016881261.1
XM_017025773.1 2852 UTR 3 XP_016881262.1
XM_017025774.1 2852 UTR 3 XP_016881263.1
XM_017025775.1 2852 Intron XP_016881264.1
XM_017025776.1 2852 UTR 3 XP_016881265.1
XM_017025777.1 2852 Intron XP_016881266.1

View Full Product Details