Product Details

SNP ID
rs188960721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41425900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGCCACCTTCGAAGAAGGACTC[A/G]GGCACATAGAAGGGTCCTCCTGGCT
Phenotype
MIM: 615357 MIM: 608348
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5SL PubMed Links

Gene Details

Gene
ATP5SL
Gene Name
ATP5S like
There are no transcripts associated with this gene.

Gene
B3GNT8
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198540.2 2652 Silent Mutation CCC,CCT P293P NP_940942.1
XM_011526934.2 2652 Silent Mutation CCC,CCT P293P XP_011525236.1
Gene
BCKDHA
Gene Name
branched chain keto acid dehydrogenase E1, alpha polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000709.3 2652 Intron NP_000700.1
NM_001164783.1 2652 Intron NP_001158255.1

View Full Product Details