Product Details

SNP ID
rs189551861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52155110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAATACATTTGTATGGCTTCTCT[C/T]CAGTGTGATTTCTTTGATGGTACAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF836 PubMed Links

Gene Details

Gene
ZNF836
Gene Name
zinc finger protein 836
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102657.1 2982 Missense Mutation GAA,GGA E858G NP_001096127.1
XM_011526558.2 2982 Missense Mutation GAA,GGA E858G XP_011524860.1
XM_011526559.2 2982 Missense Mutation GAA,GGA E858G XP_011524861.1
XM_017026389.1 2982 Intron XP_016881878.1

View Full Product Details