Product Details

SNP ID
rs189730184
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:145937670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATCGGAAGGCACCAATCCCTGT[T/A]TTTGGGTTGTCAGCTTCCTCCCTGT
Phenotype
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
LIX1L PubMed Links

Gene Details

Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153713.2 719 Missense Mutation AAA,AAT K209N NP_714924.1
XM_017000316.1 719 Intron XP_016855805.1

View Full Product Details