Product Details

SNP ID
rs190098119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8693995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGCTTGGAGGATAGGATCTGT[G/T]GGGGCAGAGGAGCCACAGCAGCCCA
Phenotype
MIM: 603637 MIM: 611333 MIM: 140750
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RPL27A PubMed Links

Gene Details

Gene
RPL27A
Gene Name
ribosomal protein L27a
There are no transcripts associated with this gene.

Gene
SNORA3A
Gene Name
small nucleolar RNA, H/ACA box 3A
There are no transcripts associated with this gene.

Gene
SNORA3B
Gene Name
small nucleolar RNA, H/ACA box 3B
There are no transcripts associated with this gene.

Gene
ST5
Gene Name
suppression of tumorigenicity 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005418.3 3909 UTR 3 NP_005409.3
NM_139157.2 3909 UTR 3 NP_631896.1
NM_213618.1 3909 UTR 3 NP_998783.1
XM_005253077.1 3909 UTR 3 XP_005253134.1
XM_005253083.2 3909 UTR 3 XP_005253140.1
XM_011520309.1 3909 UTR 3 XP_011518611.1
XM_011520310.1 3909 UTR 3 XP_011518612.1
XM_011520311.2 3909 UTR 3 XP_011518613.1
XM_011520312.1 3909 UTR 3 XP_011518614.1
XM_011520313.1 3909 UTR 3 XP_011518615.1
XM_011520314.1 3909 UTR 3 XP_011518616.1
XM_011520315.1 3909 UTR 3 XP_011518617.1
XM_011520316.1 3909 UTR 3 XP_011518618.1
XM_011520317.1 3909 UTR 3 XP_011518619.1
XM_011520318.1 3909 UTR 3 XP_011518620.1
XM_011520319.2 3909 UTR 3 XP_011518621.1
XM_011520320.1 3909 Intron XP_011518622.1
XM_011520321.2 3909 Intron XP_011518623.1
XM_011520322.1 3909 Intron XP_011518624.1
XM_011520323.2 3909 UTR 3 XP_011518625.2
XM_011520324.2 3909 UTR 3 XP_011518626.2
XM_011520325.1 3909 UTR 3 XP_011518627.1
XM_011520326.1 3909 UTR 3 XP_011518628.1
XM_011520327.1 3909 UTR 3 XP_011518629.1
XM_011520328.1 3909 UTR 3 XP_011518630.1
XM_011520329.1 3909 UTR 3 XP_011518631.1
XM_017018182.1 3909 UTR 3 XP_016873671.1
XM_017018183.1 3909 UTR 3 XP_016873672.1
XM_017018184.1 3909 UTR 3 XP_016873673.1
XM_017018185.1 3909 UTR 3 XP_016873674.1
XM_017018186.1 3909 UTR 3 XP_016873675.1
XM_017018187.1 3909 UTR 3 XP_016873676.1

View Full Product Details