Product Details

SNP ID
rs190357072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184089229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTTTTAAAACACATTACATAAA[T/C]CTTTAGAAATCTTTTGTGAAGGTAA
Phenotype
MIM: 616983
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ENPP6 PubMed Links

Gene Details

Gene
ENPP6
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153343.3 3413 UTR 3 NP_699174.1

View Full Product Details