Product Details

SNP ID
rs191012228
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:68476975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGTTTTCTTAAAATCAGCATT[C/G]GTTGCTTGATTGTAGGTTTGTCACC
Phenotype
MIM: 612002
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DEPDC1 PubMed Links

Gene Details

Gene
DEPDC1
Gene Name
DEP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114120.2 2615 Missense Mutation CCA,CGA P798R NP_001107592.1
NM_017779.5 2615 Missense Mutation CCA,CGA P514R NP_060249.2

View Full Product Details