Product Details

SNP ID
rs190708585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30635397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCTATGGCCGTGGCTATGGTG[C/G]CTATGGATATGGCTGCTGCCGCCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP20-2 PubMed Links

Gene Details

Gene
KRTAP20-2
Gene Name
keratin associated protein 20-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181616.1 134 Missense Mutation GCC,GGC A45G NP_853647.1
Gene
KRTAP20-3
Gene Name
keratin associated protein 20-3
There are no transcripts associated with this gene.

View Full Product Details