Product Details

SNP ID
rs190673193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5105788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTCCTAAAGTAGATTAGACCGA[T/C]GTAGAATTCTGTCTGGAGACGTTCT
Phenotype
MIM: 194480 MIM: 616463
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
ZFP3
Gene Name
ZFP3 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 1489 UTR 3 NP_001307881.1
NM_001320953.1 1489 UTR 3 NP_001307882.1
NM_001320954.1 1489 UTR 3 NP_001307883.1
NM_001320955.1 1489 UTR 3 NP_001307884.1
NM_014519.3 1489 UTR 3 NP_055334.2
XM_011524006.2 1489 UTR 3 XP_011522308.1
XM_017025021.1 1489 UTR 3 XP_016880510.1
XM_017025022.1 1489 UTR 3 XP_016880511.1
XM_017025023.1 1489 UTR 3 XP_016880512.1

View Full Product Details