Product Details

SNP ID
rs191594540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:86531290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATAAGGCACTGGGCAAATGTGAC[G/A]TAGGCTGGGATGAAACCCATTCTCC
Phenotype
MIM: 616820
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MTHFSD PubMed Links

Gene Details

Gene
MTHFSD
Gene Name
methenyltetrahydrofolate synthetase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159377.1 1924 UTR 3 NP_001152849.1
NM_001159378.1 1924 UTR 3 NP_001152850.1
NM_001159379.1 1924 UTR 3 NP_001152851.1
NM_001159380.1 1924 UTR 3 NP_001152852.1
NM_022764.2 1924 UTR 3 NP_073601.2
XM_005256101.1 1924 UTR 3 XP_005256158.1
XM_005256105.1 1924 UTR 3 XP_005256162.1
XM_005256106.1 1924 UTR 3 XP_005256163.1
XM_006721247.3 1924 UTR 3 XP_006721310.1
XM_011523280.2 1924 UTR 3 XP_011521582.1
XM_011523282.2 1924 Intron XP_011521584.1
XM_011523283.2 1924 Intron XP_011521585.1
XM_011523284.2 1924 Intron XP_011521586.1
XM_011523285.1 1924 UTR 3 XP_011521587.1
XM_011523286.1 1924 UTR 3 XP_011521588.1
XM_011523287.1 1924 UTR 3 XP_011521589.1
XM_011523288.1 1924 UTR 3 XP_011521590.1
XM_011523289.1 1924 UTR 3 XP_011521591.1
XM_017023571.1 1924 UTR 3 XP_016879060.1
XM_017023572.1 1924 UTR 3 XP_016879061.1
XM_017023573.1 1924 UTR 3 XP_016879062.1
XM_017023574.1 1924 UTR 3 XP_016879063.1

View Full Product Details