Product Details

SNP ID
rs191924906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:81991388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAATCCCATCTGCTTTACCACC[A/G]GAAGATTGCACTGTGACTCAGTCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATA31D1 PubMed Links

Gene Details

Gene
SPATA31D1
Gene Name
SPATA31 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001670.2 943 Silent Mutation CCA,CCG P306P NP_001001670.1
XM_017014710.1 943 Silent Mutation CCA,CCG P269P XP_016870199.1

View Full Product Details