Product Details

SNP ID
rs191651536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25881112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTTAATTCCTATATCACAAAT[A/G]TAGCAGAAGCAGCAATCTGTACAGT
Phenotype
MIM: 104760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APP PubMed Links

Gene Details

Gene
APP
Gene Name
amyloid beta precursor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000484.3 2960 UTR 3 NP_000475.1
NM_001136016.3 2960 UTR 3 NP_001129488.1
NM_001136129.2 2960 UTR 3 NP_001129601.1
NM_001136130.2 2960 UTR 3 NP_001129602.1
NM_001136131.2 2960 UTR 3 NP_001129603.1
NM_001204301.1 2960 UTR 3 NP_001191230.1
NM_001204302.1 2960 UTR 3 NP_001191231.1
NM_001204303.1 2960 UTR 3 NP_001191232.1
NM_201413.2 2960 UTR 3 NP_958816.1
NM_201414.2 2960 UTR 3 NP_958817.1

View Full Product Details