Product Details

SNP ID
rs192551710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59713028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTGTAGGCCACAGATACCACC[C/T]GGCCCATCTCAGGAGAGTAGCTGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10V1 PubMed Links

Gene Details

Gene
OR10V1
Gene Name
olfactory receptor family 10 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005324.1 818 Missense Mutation CAG,CGG Q273R NP_001005324.1

View Full Product Details