Product Details
- SNP ID
-
rs192230740
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:27879509 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTATACTGCAAGACAGAGAGATT[C/T]GAAGTGGAGGAGGGAAGCCAACAGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LYRM9
PubMed Links
Gene Details
- Gene
- LYRM9
- Gene Name
- LYR motif containing 9
View Full Product Details