Product Details

SNP ID
rs193118599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5967915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTAGCTCTGATTCAAGGTGATGT[C/T]ATCACAAGAGAGTTTAGACACAGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR56A5 PubMed Links

Gene Details

Gene
OR56A5
Gene Name
olfactory receptor family 56 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146033.1 580 Missense Mutation AAC,GAC N194D NP_001139505.1

View Full Product Details