Product Details

SNP ID
rs193219264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119044197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACAGCACAAAAACTGTAGAAAA[G/T]AATATTTTATTGAAACAGTTTCTCA
Phenotype
MIM: 601746
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HYOU1 PubMed Links

Gene Details

Gene
HYOU1
Gene Name
hypoxia up-regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130991.2 4494 UTR 3 NP_001124463.1
NM_006389.4 4494 UTR 3 NP_006380.1
XM_005271392.3 4494 UTR 3 XP_005271449.1
XM_005271393.3 4494 UTR 3 XP_005271450.1
XM_005271394.3 4494 UTR 3 XP_005271451.1
XM_017017095.1 4494 UTR 3 XP_016872584.1
XM_017017096.1 4494 UTR 3 XP_016872585.1
XM_017017097.1 4494 UTR 3 XP_016872586.1

View Full Product Details