Product Details

SNP ID
rs193279150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19711283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGTATTCACACAGCTTCTGTCC[A/G]GTATGAACTCTTTTGTGTATTCAGA
Phenotype
MIM: 194556
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF14 PubMed Links

Gene Details

Gene
ZNF14
Gene Name
zinc finger protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021030.2 2152 UTR 3 NP_066358.2

View Full Product Details