Product Details
- SNP ID
-
rs6051581
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:3109323 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCATGAGGAAACGCTGCGCTCCCC[A/C]TCAGGGAGCAGTTTCTGAAGCCAGC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UBOX5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142019808] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UBOX5
- Gene Name
- U-box domain containing 5
- Gene
- UBOX5-AS1
- Gene Name
- UBOX5 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details