Product Details
- SNP ID
-
rs7692418
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:78901071 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTTTTCCTTTTTTGAGATAAGAGT[C/T]TTGCTCTGTTGCCCAGGCTGGAGTG
- Phenotype
-
MIM: 614577
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BMP2K
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7692450] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BMP2K
- Gene Name
- BMP2 inducible kinase
- Gene
- PAQR3
- Gene Name
- progestin and adipoQ receptor family member 3
View Full Product Details