Product Details

SNP ID
rs7893052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:12752257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACTATATACCTCTTCATTGATT[C/T]AGACCTTTGAATTATTTATTCACAC
Phenotype
MIM: 400005
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP9Y PubMed Links

Gene Details

Gene
USP9Y
Gene Name
ubiquitin specific peptidase 9, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004654.3 Intron NP_004645.2
XM_017030078.1 Intron XP_016885567.1

View Full Product Details