Product Details
- SNP ID
-
rs200995685
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:116664837 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTCCTTGTAGCAGTTTTTGGCCA[C/T]GACAGTGTATCTCGTGCCACAGCCA
- Phenotype
-
MIM: 610701
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C10orf82
PubMed Links
Gene Details
- Gene
- C10orf82
- Gene Name
- chromosome 10 open reading frame 82
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144661.2 |
858 |
Missense Mutation |
ATG,GTG |
M129V |
NP_653262.1 |
XM_011539340.2 |
858 |
Missense Mutation |
ATG,GTG |
M152V |
XP_011537642.1 |
XM_011539341.1 |
858 |
Missense Mutation |
ATG,GTG |
M129V |
XP_011537643.1 |
XM_011539342.2 |
858 |
Missense Mutation |
ATG,GTG |
M129V |
XP_011537644.1 |
XM_011539343.2 |
858 |
Missense Mutation |
ATG,GTG |
M152V |
XP_011537645.1 |
XM_011539344.2 |
858 |
Missense Mutation |
ATG,GTG |
M152V |
XP_011537646.1 |
XM_011539345.2 |
858 |
Missense Mutation |
ATG,GTG |
M152V |
XP_011537647.1 |
XM_011539346.2 |
858 |
Missense Mutation |
ATG,GTG |
M152V |
XP_011537648.1 |
XM_011539347.2 |
858 |
Missense Mutation |
ATG,GTG |
M152V |
XP_011537649.1 |
XM_017015753.1 |
858 |
Missense Mutation |
ATG,GTG |
M129V |
XP_016871242.1 |
XM_017015754.1 |
858 |
Missense Mutation |
ATG,GTG |
M129V |
XP_016871243.1 |
XM_017015755.1 |
858 |
Missense Mutation |
ATG,GTG |
M129V |
XP_016871244.1 |
- Gene
- HSPA12A
- Gene Name
- heat shock protein family A (Hsp70) member 12A
There are no transcripts associated with this gene.
- Gene
- LOC105378499
- Gene Name
- uncharacterized LOC105378499
There are no transcripts associated with this gene.
View Full Product Details