Product Details
- SNP ID
-
rs200632520
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:71712706 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGCGACACACGGGCACAGCCACC[A/G]TGTTCGTCACTGTCCTGGATGTGAA
- Phenotype
-
MIM: 605516
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C10orf105
PubMed Links
Gene Details
- Gene
- C10orf105
- Gene Name
- chromosome 10 open reading frame 105
- Gene
- CDH23
- Gene Name
- cadherin-related 23
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001171930.1 |
3832 |
Missense Mutation |
ATG,GTG |
M1088V |
NP_001165401.1 |
NM_001171931.1 |
3832 |
Intron |
|
|
NP_001165402.1 |
NM_001171932.1 |
3832 |
Intron |
|
|
NP_001165403.1 |
NM_001171933.1 |
3832 |
Intron |
|
|
NP_001165404.1 |
NM_001171934.1 |
3832 |
Intron |
|
|
NP_001165405.1 |
NM_001171935.1 |
3832 |
Intron |
|
|
NP_001165406.1 |
NM_001171936.1 |
3832 |
Intron |
|
|
NP_001165407.1 |
NM_022124.5 |
3832 |
Missense Mutation |
ATG,GTG |
M1088V |
NP_071407.4 |
NM_052836.3 |
3832 |
Intron |
|
|
NP_443068.1 |
XM_006717940.3 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_006718003.1 |
XM_006717942.3 |
3832 |
Missense Mutation |
ATG,GTG |
M1131V |
XP_006718005.1 |
XM_011540039.2 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_011538341.1 |
XM_011540042.2 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_011538344.1 |
XM_011540043.2 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_011538345.1 |
XM_011540044.2 |
3832 |
Missense Mutation |
ATG,GTG |
M1108V |
XP_011538346.1 |
XM_011540045.2 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_011538347.1 |
XM_011540046.2 |
3832 |
Missense Mutation |
ATG,GTG |
M973V |
XP_011538348.1 |
XM_011540047.2 |
3832 |
Missense Mutation |
ATG,GTG |
M759V |
XP_011538349.1 |
XM_011540048.2 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_011538350.1 |
XM_011540049.2 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_011538351.1 |
XM_011540051.2 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_011538353.1 |
XM_011540052.2 |
3832 |
Intron |
|
|
XP_011538354.1 |
XM_017016499.1 |
3832 |
Missense Mutation |
ATG,GTG |
M1133V |
XP_016871988.1 |
XM_017016500.1 |
3832 |
Missense Mutation |
ATG,GTG |
M1153V |
XP_016871989.1 |
XM_017016501.1 |
3832 |
Missense Mutation |
ATG,GTG |
M299V |
XP_016871990.1 |
XM_017016502.1 |
3832 |
Missense Mutation |
ATG,GTG |
M299V |
XP_016871991.1 |
XM_017016503.1 |
3832 |
Intron |
|
|
XP_016871992.1 |
XM_017016504.1 |
3832 |
Intron |
|
|
XP_016871993.1 |
XM_017016505.1 |
3832 |
Intron |
|
|
XP_016871994.1 |
XM_017016506.1 |
3832 |
Intron |
|
|
XP_016871995.1 |
XM_017016507.1 |
3832 |
Intron |
|
|
XP_016871996.1 |
XM_017016508.1 |
3832 |
Intron |
|
|
XP_016871997.1 |
View Full Product Details