Product Details

SNP ID
rs200778412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114833249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTTTAGATGATAAAGCCCCAGT[C/G]ACCGATACAAATATTCCATCGCATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM160B1 PubMed Links

Gene Details

Gene
FAM160B1
Gene Name
family with sequence similarity 160 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135051.1 476 Silent Mutation GTC,GTG V47V NP_001128523.1
NM_020940.3 476 Silent Mutation GTC,GTG V47V NP_065991.3

View Full Product Details