Product Details

SNP ID
rs200173023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70771439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCTGGCTTGAATGCATCTGGAG[A/G]ACTTGAGCTTCTCCAATGTACTCTA
Phenotype
MIM: 607506 MIM: 612640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS14 PubMed Links

Gene Details

Gene
ADAMTS14
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
There are no transcripts associated with this gene.

Gene
TBATA
Gene Name
thymus, brain and testes associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318241.1 1632 Silent Mutation GTC,GTT V332V NP_001305170.1
NM_001318242.1 1632 Silent Mutation GTC,GTT V331V NP_001305171.1
NM_001318243.1 1632 Silent Mutation GTC,GTT V330V NP_001305172.1
NM_152710.3 1632 Silent Mutation GTC,GTT V331V NP_689923.3
XM_017015845.1 1632 UTR 3 XP_016871334.1
XM_017015846.1 1632 UTR 3 XP_016871335.1
XM_017015847.1 1632 UTR 3 XP_016871336.1
XM_017015848.1 1632 Missense Mutation CCT,TCT P380S XP_016871337.1
XM_017015849.1 1632 Missense Mutation TCC,TTC S395F XP_016871338.1
XM_017015850.1 1632 Intron XP_016871339.1
XM_017015851.1 1632 Intron XP_016871340.1
XM_017015852.1 1632 Missense Mutation CCT,TCT P346S XP_016871341.1
XM_017015853.1 1632 Silent Mutation GTC,GTT V363V XP_016871342.1
XM_017015854.1 1632 Intron XP_016871343.1
XM_017015855.1 1632 Intron XP_016871344.1
XM_017015856.1 1632 UTR 3 XP_016871345.1
XM_017015857.1 1632 Intron XP_016871346.1
XM_017015858.1 1632 UTR 3 XP_016871347.1
XM_017015859.1 1632 Intron XP_016871348.1
XM_017015860.1 1632 Missense Mutation CCT,TCT P288S XP_016871349.1
XM_017015861.1 1632 Intron XP_016871350.1
XM_017015862.1 1632 Intron XP_016871351.1

View Full Product Details