Product Details

SNP ID
rs200014503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55665314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTATTGTGGTAACTATTATCACA[A/G]GTCAGAGTCTGAGGTCACCTATGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4C6 PubMed Links

Gene Details

Gene
OR4C6
Gene Name
olfactory receptor family 4 subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004704.1 148 Missense Mutation AGT,GGT S50G NP_001004704.1

View Full Product Details