Product Details

SNP ID
rs201960565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1697661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTGACTTTGGACCTCAGGTGA[G/C]TCCAGCATATCCACACCACCCAAGA
Phenotype
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP5-6 PubMed Links

Gene Details

Gene
KRTAP5-6
Gene Name
keratin associated protein 5-6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012416.1 467 UTR 3 NP_001012416.1

View Full Product Details