Product Details

SNP ID
rs201404581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1630536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCAGCTGCTGTGTCCCCGTGTG[C/G]TGCCAGTGTAAGATCTGAGGCTCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP5-5 PubMed Links

Gene Details

Gene
KRTAP5-5
Gene Name
keratin associated protein 5-5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001480.2 734 Missense Mutation TGC,TGG C232W NP_001001480.2

View Full Product Details