Product Details

SNP ID
rs199726391
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56361316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCTGAACTTACTACTTCTGAT[A/G]TGAAAGAAGCCAGAGTAGTTGTTTC
Phenotype
MIM: 107760 MIM: 600556
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
APOF PubMed Links

Gene Details

Gene
APOF
Gene Name
apolipoprotein F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001638.2 944 Missense Mutation ACA,ATA T297I NP_001629.1
Gene
STAT2
Gene Name
signal transducer and activator of transcription 2
There are no transcripts associated with this gene.

View Full Product Details