Product Details

SNP ID
rs201742326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50331732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTAGTAATTTTAAGAATGGCTT[C/T]TGAACAAATGTGGTAGAGGCAGGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM186A PubMed Links

Gene Details

Gene
FAM186A
Gene Name
family with sequence similarity 186 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145475.1 6923 Silent Mutation CAA,CAG Q2262Q NP_001138947.1
XM_006719231.3 6923 Silent Mutation CAA,CAG Q2262Q XP_006719294.1
XM_011537890.2 6923 Silent Mutation CAA,CAG Q2258Q XP_011536192.1
XM_011537892.1 6923 Silent Mutation CAA,CAG Q2119Q XP_011536194.1

View Full Product Details