Product Details

SNP ID
rs201962193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14805273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGTTTTGCTTTCTTGTCTTCT[A/-]AATGTTTCAGTGTTTGAACATTCAC
Phenotype
Polymorphism
A/-, Insertion/deletion
Allele Nomenclature
Literature Links
C12orf60 PubMed Links

Gene Details

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175874.3 4146 Intron NP_787070.2
XM_005253322.4 4146 Intron XP_005253379.1
XM_011520568.2 4146 Intron XP_011518870.1
XM_011520569.2 4146 Intron XP_011518871.1
XM_017018871.1 4146 Intron XP_016874360.1
XM_017018872.1 4146 Intron XP_016874361.1
XM_017018873.1 4146 Intron XP_016874362.1
XM_017018874.1 4146 Intron XP_016874363.1
Gene
SMCO3
Gene Name
single-pass membrane protein with coiled-coil domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013698.2 4146 UTR 3 NP_001013720.2
XM_017019312.1 4146 UTR 3 XP_016874801.1
Gene
WBP11
Gene Name
WW domain binding protein 11
There are no transcripts associated with this gene.

View Full Product Details