Product Details
- SNP ID
-
rs201311103
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:77051091 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACAAAAATGTGTTGTTCCAAAACA[A/G]AAAAAAACAGCCACTGAACAGCAGT
- Phenotype
-
MIM: 610392
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYCBP2
PubMed Links
Gene Details
- Gene
- MYCBP2
- Gene Name
- MYC binding protein 2, E3 ubiquitin protein ligase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015057.4 |
13962 |
Silent Mutation |
TTC,TTT |
F4609F |
NP_055872.4 |
XM_005266299.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4686F |
XP_005266356.1 |
XM_006719779.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4728F |
XP_006719842.1 |
XM_006719780.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4725F |
XP_006719843.1 |
XM_006719781.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4720F |
XP_006719844.1 |
XM_006719782.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4703F |
XP_006719845.1 |
XM_006719783.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4694F |
XP_006719846.1 |
XM_006719784.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4692F |
XP_006719847.1 |
XM_006719785.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4668F |
XP_006719848.1 |
XM_006719786.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4634F |
XP_006719849.1 |
XM_006719787.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4628F |
XP_006719850.1 |
XM_006719788.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4620F |
XP_006719851.1 |
XM_006719789.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4683F |
XP_006719852.1 |
XM_011535004.2 |
13962 |
Silent Mutation |
TTC,TTT |
F4619F |
XP_011533306.1 |
XM_011535005.2 |
13962 |
Silent Mutation |
TTC,TTT |
F3773F |
XP_011533307.1 |
XM_017020454.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4709F |
XP_016875943.1 |
XM_017020455.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4684F |
XP_016875944.1 |
XM_017020456.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4667F |
XP_016875945.1 |
XM_017020457.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4661F |
XP_016875946.1 |
XM_017020458.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4650F |
XP_016875947.1 |
XM_017020459.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4598F |
XP_016875948.1 |
XM_017020460.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4594F |
XP_016875949.1 |
XM_017020461.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4590F |
XP_016875950.1 |
XM_017020462.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4586F |
XP_016875951.1 |
XM_017020463.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4573F |
XP_016875952.1 |
XM_017020464.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4568F |
XP_016875953.1 |
XM_017020465.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4550F |
XP_016875954.1 |
XM_017020466.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4534F |
XP_016875955.1 |
XM_017020467.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4531F |
XP_016875956.1 |
XM_017020468.1 |
13962 |
Silent Mutation |
TTC,TTT |
F4463F |
XP_016875957.1 |
View Full Product Details