Product Details

SNP ID
rs200221043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39038702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTTGCATTTTTGGTTTTGCA[C/T]TCGCGTTTTTGTGGCTCTCCAGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NHLRC3 PubMed Links

Gene Details

Gene
NHLRC3
Gene Name
NHL repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012754.3 392 Silent Mutation CAC,CAT H21H NP_001012772.1
NM_001017370.2 392 Silent Mutation CAC,CAT H21H NP_001017370.1
Gene
PROSER1
Gene Name
proline and serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025138.4 392 Intron NP_079414.3
XM_005266545.3 392 Intron XP_005266602.1
XM_011535239.2 392 Intron XP_011533541.1

View Full Product Details