Product Details

SNP ID
rs200012590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20590405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGCCTTCAATGAGCTGACAGA[A/C]TGTCATTTTGAACTTTGTCTCACTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC254028 PubMed Links

Gene Details

Gene
LOC254028
Gene Name
uncharacterized LOC254028
There are no transcripts associated with this gene.

Gene
RNASE11
Gene Name
ribonuclease A family member 11 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145250.3 419 Intron NP_660293.1
Gene
RNASE12
Gene Name
ribonuclease A family member 12 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024822.2 419 Missense Mutation GTC,TTC V107F NP_001019993.1

View Full Product Details