Product Details

SNP ID
hCV190122974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41349106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACCTCTCTTTTCAGGATGAAAG[C/T]CAAACTTCTGCATCCTCTTGTGATG
Phenotype
MIM: 612818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUSAP1 PubMed Links

Gene Details

Gene
NUSAP1
Gene Name
nucleolar and spindle associated protein 1
There are no transcripts associated with this gene.

View Full Product Details