Product Details
- SNP ID
-
rs201269482
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:66337059 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTCCTCTTCATTTAAAGTATTGG[C/T]AATAGCATCATTACATGGATTGTCC
- Phenotype
-
MIM: 614183
MIM: 610716
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DIS3L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34848112,rs3759786] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DIS3L
- Gene Name
- DIS3 like exosome 3'-5' exoribonuclease
There are no transcripts associated with this gene.
- Gene
- TIPIN
- Gene Name
- TIMELESS interacting protein
View Full Product Details